Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPSPGTVCSL LLLGMLWLDL AMAGSSFLSP EHQRVQQRKE SKKPPAKLQP RALAGWLRPE DGGQAEGAED EMEVRFNAPF DVGIKLSGVQ YQQHSQALGK FLQDILWEEA KEAPADK
GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMCBX1; Appetite-regulating hormone; Fip3p; ghrelin 27; ghrelin 28; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; Growth hormone secretagogue; Growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP1; IP2; IPD2; Motilin-related peptide; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; Protein M46
Gene Aliases: GHRL; MTLRP; UNQ524/PRO1066
UniProt ID: (Human) A8CF34
Entrez Gene ID: (Human) 51738
Molecular Function: peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support