Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
May cause the redistribution of PAPOLB from the cytosol to the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Germ cell-specific gene 1 protein; testicular secretory protein Li 20
Gene Aliases: GSG1; UNQ709/PRO1360
UniProt ID: (Human) Q2KHT4
Entrez Gene ID: (Human) 83445
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support