Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MANEVIKCK AAVAWEAGKP LSIEEIEVAP PKAHEVRIKI IATAVCHTDA YTLSGADPEG CFPVILGHEG AGIVESVGEG VTKLKAGDTV IPLYIPQCGE CKFCLNPKTN LCQKIRVTQG KGLMPDGTSR FTCKGKTILH YMGTSTFSEY TVVADISVAK IDPLAPLDKV CLLGCGISTG YGAAVNTAKL EPGSVCAVFG LGGVGLAVIM GCKVAGASRI IGVDINKDKF ARAKEFGATE CINPQDFSKP IQEVLIEMTD GGVDYSFECI GNVKVMRAAL EACHKGWGVS VVVGVAASGE EIATRPFQLV TGRTWKGTAF GGWKSVESVP KLVSEYMSKK IKVDEFVTHN LSFDEINKAF ELMHSGKSIR TVVKI (1-374 aa encoded by BC014665)
This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADH-B2; alcohol dehydrogenase (class III), chi polypeptide; Alcohol dehydrogenase 2; Alcohol dehydrogenase 5; Alcohol dehydrogenase B2; Alcohol dehydrogenase class chi chain; Alcohol dehydrogenase class-3; Alcohol dehydrogenase class-III; class III alcohol dehydrogenase, chi subunit; epididymis secretory sperm binding protein Li 60p; FALDH; FDH; formaldehyde dehydrogenase; Glutathione-dependent formaldehyde dehydrogenase; GSH-FDH; S-(hydroxymethyl)glutathione dehydrogenase; S-nitrosoglutathione reductase
Gene Aliases: Adh-2; ADH-3; Adh-5; Adh2; Adh3; ADH5; ADHX; FALDH; FDH; GSH-FDH; GSNOR; HEL-S-60p
UniProt ID: (Human) P11766, (Rat) P12711, (Mouse) P28474
Entrez Gene ID: (Human) 128, (Pig) 100513555, (Rat) 100145871, (Mouse) 11532
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support