Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human CTH.
Immunogen sequence: AVAALDGAK YCLAFASGLA ATVTITHLLK AGDQIICMDD VYGGTNRYFR QVASEFGLKI SFVDCSKIKL LEAAITPETK LVWIETPTNP TQKVIDIEGC AHIVHKHGDI ILVVDNTFMS PYFQRPLALG ADISMYSATK YMNGHSDVVM GLVSVNCESL HNRLRFLQNS LGAVPSPIDC YLCNRGLKTL HVRMEKHFKN GMAVAQFLES NPWVEKVIYP GLPSHPQHEL VKRQCTGCTG MVTFYIKGTL QHAEIFLKNL KLFTLAESLG GFESLAELPA IMTHASVLKN DRDVLGISDT LIRLSVGLED EEDLLEDLDQ ALKAAHPPSG SHS (74-406 aa encoded by BC015807)
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTL target antigen; cystathionase (cystathionine gamma-lyase); cysteine desulfhydrase; cysteine-protein sulfhydrase; gamma-cystathionase; homocysteine desulfhydrase; homoserine deaminase; homoserine dehydratase; MGC9471; PRB-RA; probasin-related antigen; unnamed protein product
Gene Aliases: 0610010I13Rik; CGL; CSE; Cys3
UniProt ID: (Human) P32929, (Rat) P18757, (Mouse) Q8VCN5
Entrez Gene ID: (Human) 1491, (Rat) 24962, (Mouse) 107869
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support