Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVSKLTSLQQ ELLSALLSSG VTKEVLVQAL EELLPSPNFG VKLETLPLSP GSGAEPDTKP VFHTLTNGHA KGRLSGDEGS EDGDDYDTPP ILKELQALNT EEAAEQRAEV DRMLSEDPWR AAKMIKGYMQ QHNIPQREVV DVTGLNQSHL SQHLNKGTPM KTQKRAALYT WYVRKQREIL RQFNQTVQSS GNMTDKSSQD QLLFLFPEFS QQSHGPGQSD DACSEPTNKK MRRNRFKWGP ASQQILYQAY DRQKNPSKEE REALVEECNR AECLQRGVSP SKAHGLGSNL VTEVRVYNWF ANRRKEEAFR QKLAMDAYSS NQTHSLNPLL SHGSPHHQPS SSPPNKLSGV RYSQQGNNEI TSSSTISHHG NSAMVTSQSV LQQVSPASLD PGHNLLSPDG KMISVSGGGL PPVSTLTNIH SLSHHNPQQS QNLIMTPLSG VMAIAQSLNT SQAQSVPVIN SVAGSLAALQ PVQFSQQLHS PHQQPLMQQS PGSHMAQQPF MAAVTQLQNS HMYAHKQEPP QYSHTSRFPS AMVVTDTSSI STLTNMSSSK QCPLQAW
Hepatocyte Nuclear Factor 1 (HNF1, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor) is a member of the homeodomain-containing superfamily of transcription factors. It is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. HNF1has been shown to function in nephron development and regulates development of the embryonic pancreas. Mutations in the gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of the gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocyte nuclear factor 1-beta; HNF-1-beta; HNF1 beta A; Homeoprotein LFB3; TCF-2; Transcription factor 2; transcription factor 2, hepatic; Variant hepatic nuclear factor 1; vHNF1
Gene Aliases: FJHN; HNF-1-beta; HNF-1B; HNF1B; HNF1beta; HNF2; HPC11; LF-B3; LFB3; MODY5; TCF-2; TCF2; VHNF1
UniProt ID: (Human) P35680
Entrez Gene ID: (Human) 6928
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support