Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with mouse, pig and cow based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factorm HOXA10, that may regulate gene expression, morphogenesis, and differentiation. More specifically, HOXA10 may function in fertility, embryo viability, and regulation of hematopoietic lineage commitment. Alternatively spliced transcript variants have been described. Read-through transcription also exists between this gene and the downstream homeobox A9 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box A10; homeobox protein 1H; Homeobox protein Hox-1.8; Homeobox protein Hox-1H; Homeobox protein Hox-A10; homeobox protein HOXA10; Hox-18; PL
Gene Aliases: HOX1; HOX1.8; HOX1H; HOXA10; PL
UniProt ID: (Human) P31260
Entrez Gene ID: (Human) 3206
Molecular Function: helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support