Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: PAQPQIYPWM RKLHISHDNI GGPEGKRART AYTRYQTLEL EKEFHFNRYL TRRRRIEIAH ALCLSERQIK IWFQNRRMKW KKDNKLKSMS MAAAGGAFRP
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 1C; homeo box A5; Homeobox protein Hox-1C; Homeobox protein Hox-A5; homeobox protein HOXA5; Hox-1C
Gene Aliases: HOX1; HOX1.3; HOX1C; HOXA5
UniProt ID: (Human) P20719
Entrez Gene ID: (Human) 3202
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support