Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATTGALGNY YVDSFLLGAD AADELSVGRY APGTLGQPPR QAATLAEHPD FSPCSFQSKA TVFGASWNPV HAAGANAVPA AVYHHHHHHP YVHPQAPVAA AAPDGRYMRS WLEPTPGALS FAGLPSSRPY GIKPEPLSAR RGDCPTLDTH TLSLTDYACG SPPVDREKQP SEGAFSENNA ENESGGDKPP IDPNNPAANW LHARSTRKKR CPYTKHQTLE LEKEFLFNMY LTRDRRYEVA RLLNLTERQV KIWFQNRRMK MKKINKDRAK DE
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein Hox-1G; Homeobox protein Hox-A9; homeodomain protein HOXA9; HOXA 9
Gene Aliases: ABD-B; HOX1; HOX1.7; HOX1G; HOXA9
UniProt ID: (Human) P31269
Entrez Gene ID: (Human) 3205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support