Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: C6, SH-SY5Y, U-87MG, Mouse testis, Mouse brain, Mouse spleen, Rat testis, Rat brain
Immunogen sequence: MMDVNSSGRP DLYGHLRSFL LPEVGRGLPD LSPDGGADPV AGSWAPHLLS EVTASPAPTW DAPPDNASGC GEQINYGRVE KVVIGSILTL ITLLTIAGNC
5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa. 5-HT7 Receptor has been reported in aorta, brain, heart, placenta, small intestine, spleen, artery, and vessel. An EST has been isolated from a testis library.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-HT-7; 5-HT-X; 5-HT7; 5-HT7 receptor; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; 5HT7 Receptor; GPRFO; high affinity serotonin receptor (5HT7); HT7; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; Serotonin receptor 7
Gene Aliases: 5-HT7; 5Ht7; HTR7
UniProt ID: (Human) P34969, (Mouse) P32304, (Rat) P32305
Entrez Gene ID: (Human) 3363, (Mouse) 15566, (Rat) 65032
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support