Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-H2, rhB7-H3, rhB7-H3b, rhPD-L2, recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H3, rmB7-H4, rmPD-L2, or recombinant rat B7-2 is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B7 homolog 2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP-1; CD275; ICOS ligand; LICOS
Gene Aliases: AU044799; B7-H2; B7h; B7h2; B7RP-1; B7rp1; BG071784; GI50; GL50; GL50-B; ICOS-L; Icosl; Icoslg; LICOS; Ly115l; mKIAA0653
UniProt ID: (Mouse) Q9JHJ8
Entrez Gene ID: (Mouse) 50723
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support