Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs, no cross-reactivity with recombinant mouse IL-28 alpha is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
The protein encoded by this gene belongs to the class II cytokine receptor family. This protein forms a receptor complex with interleukine 10 receptor, beta (IL10RB). The receptor complex has been shown to interact with three closely related cytokines, including interleukin 28A (IL28A), interleukin 28B (IL28B), and interleukin 29 (IL29). The expression of all three cytokines can be induced by viral infection. The cells overexpressing this protein have been found to have enhanced responses to IL28A and IL29, but decreased response to IL28B. Three alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class II cytokine receptor CRF2/12; CRF2-12; Cytokine receptor class-II member 12; Cytokine receptor family 2 member 12; IFN-lambda receptor 1; IFN-lambda-R1; IL-28 receptor subunit alpha; IL-28R-alpha; IL-28RA; Interferon lambda receptor 1; interferon lambda, receptor 1; interferon, lambda receptor 1; interleukin 28 alpha receptor; interleukin 28 receptor A; interleukin 28 receptor, alpha (interferon, lambda receptor); interleukin or cytokine receptor 2; Interleukin-28; Interleukin-28 receptor subunit alpha; LICR2; Likely interleukin or cytokine receptor 2
Gene Aliases: CRF2/12; IFNLR; IFNLR1; IL-28R1; IL28RA; LICR2
UniProt ID: (Human) Q8IU57
Entrez Gene ID: (Human) 163702
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support