Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 10
Positive test controls include: Raji, Mouse spleen, Mouse thymus, Rat lung.
Immunogen sequence: IYQQNHMVLS RARNGMPLLK EVPRSYELLK PPPICPRDSV KVINKEGEVM DVYRCDHCRV LFLDYVMFTI HMGCHGFRDP FECNMCGYRS HDRYEFSSHI ARGEHRALLK
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IKAROS family zinc finger 3 (Aiolos); Ikaros family zinc finger protein 3; zinc finger DNA binding protein Aiolos; Zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos)
Gene Aliases: 5830411O07Rik; AIO; AIOLOS; IKZF3; Zfpn1a3; ZNFN1A3
UniProt ID: (Human) Q9UKT9, (Mouse) O08900
Entrez Gene ID: (Human) 22806, (Mouse) 22780, (Rat) 303511
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support