Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Immunogen sequence: AHFGTDETIL DGKEYDIYVS YARNAEEEEF VLLTLRGVLE NEFGYKLCIF DRDSLPGGIV TDETLSFIQK SRRLLVVLSP
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL-1R-3; IL-1R3; IL1RAcP; ILN; Interleukin; Interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain
Gene Aliases: 6430709H04Rik; AI255955; AV239853; C3orf13; IL-1RAcP; IL1R3; Il1racpb; IL1RAP
UniProt ID: (Human) Q9NPH3, (Mouse) Q61730, (Rat) Q63621
Entrez Gene ID: (Human) 3556, (Mouse) 16180, (Rat) 25466
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support