Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
AA Sequence of recombinant protein: MECHIKDKEG KAYESVLMIS IDELDKMTGT DSNCPNNEPN FFRKHVCDDT KEAAFLNRAA RKLKQFLKMN ISEEFNVHLL TVSQGTQTLV NCTSKEEKNV KEQKKNDACF LKRLLREIKT CWNKILKGSI.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-7. Anti-Murine IL-7-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Murine IL-7 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-7 (500-P57) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-7.
Western Blot: To detect Murine IL-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine IL-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P57BT-1MG will be provided as 2 x 500 µg
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IL-7; IL-7; ILN; Interleukin; Interleukin-7; Interleukin7; M-IL-7
Gene Aliases: A630026I06Rik; hlb368; Il-7; Il7
UniProt ID: (Mouse) P10168
Entrez Gene ID: (Mouse) 16196
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support