Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total IL28B.
IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytokine Zcyto22; IFN-lambda 3; IFN-lambda-3; IFN-lambda-4; IL-28B; IL-28C; ILN; interferon alpha; Interferon lambda-3; interferon, lambda 3; interferon, lambda 4; Interleukin; interleukin 28; interleukin 28B; interleukin 28C; Interleukin-28B; Interleukin-28C
Gene Aliases: IFL-1; IFNL3; IL-28B; Il28; IL28B; IL28C; INF-alpha; INF-lambda; ZCYTO22
UniProt ID: (Human) Q8IZI9, (Mouse) Q8CGK6
Entrez Gene ID: (Human) 282617, (Rat) 685840, (Mouse) 338374
Molecular Function: interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support