Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Enhances the promotion of guanine-nucleotide exchange by PSCD2 on ARF6 in a concentration-dependent manner. Target protein binds guanine nucleotide exchange factor cytohesin 2 and increases cytohesin 2 mediated formation of ADP-ribosylation factor GTP-ase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interaction protein for cytohesin exchange factors 1 Interaction protein for cytohesin exchange factors 1; Interactor protein for cytohesin exchange factors 1; phosphoinositide binding protein PIP3-E; Phosphoinositide-binding protein PIP3-E; RP3-402L9.2
Gene Aliases: IPCEF1; KIAA0403; PIP3-E
UniProt ID: (Human) Q8WWN9
Entrez Gene ID: (Human) 26034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support