Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEGKMDENE WGYHGEGNKS LVVAHAQRCV VLRFLKFPPN RKKTSEEIFQ HLQNIVDFGK NVMKEFLGEN YVHYGEVVQL PLEFVKQLCL KIQSERPESR CDKDLDTLSG YAMCLPNLTR LQTYRFAEHR PILCVEIKPK CGFIPFSSDV THEMKHKVCR YCMHQHLKVA TGKWKQISKY CPLDLYSGNK QRMHFALKSL LQEAQNNLKI FKNGELIYGC KDARSPVADW SELAHHLKPF FFPSNGLASG PHCTRAVIRE LVHVITRVLL SGSDKGRAGT LSPGLGPQGP RVCEASPFSR SLRCQGKNTP ERSGLPKGCL LYKTLQVQML DLLDIEGLYP LYNRVERYLE EFPEERKTLQ IDGPYDEAFY QKLLDLSTED DGTVAFALTK VQQYRVAMTA KDCSIMIALS PCLQDASSDQ RPVVPSSRSR FAFSVSVLDL DLKPYESIPH QYKLDGKIVN YYSKTVRAKD NAVMSTRFKE SEDCTLVLHK V
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA476B13.1 (novel protein); FLJ13163; inositol 1,3,4,5,6-pentakisphosphate 2-kinase; Inositol-1,3,4,5,6-pentakisphosphate 2-kinase; Inositol-pentakisphosphate 2-kinase; Ins(1,3,4,5,6)P5 2-kinase; InsP5 2-kinase; IPK1 homolog; KIAA0699
Gene Aliases: bA476B13.1; C9orf12; INSP5K2; IP5K; IPK1; IPPK
UniProt ID: (Human) Q9H8X2
Entrez Gene ID: (Human) 64768
Molecular Function: kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support