Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MQTFLKGKRV GYWLSEKKIK KLNFQAFAEL CRKRGMEVVQ LNLSRPIEEQ GPLDVIIHKL TDVILEADQN DSQSLELVHR FQEYIDAHPE TIVLDPLPAI RTLLDRSKSY ELIRKIEAYM EDDRICSPPF MELTSLCGDD TMRLLEKNGL TFPFICKTRV AHGTNSHEMA IVFNQEGLNA IQPPCVVQNF INHNAVLYKV FVVGESYTVV QRPSLKNFSA GTSDRESIFF NSHNVSKPES SSVLTELDKI EGVFERPSDE VIRELSRALR QALGVSLFGI DIIINNQTGQ HAVIDINAFP GDCQVCFIEG WKTD
ITPK1 encodes an enzyme that belongs to the inositol 1,3,4-trisphosphate 5/6-kinase family. This enzyme regulates the synthesis of inositol tetraphosphate, and downstream products, inositol pentakisphosphate and inositol hexakisphosphate. Inositol metabolism plays a role in the development of the neural tube. Disruptions in this gene are thought to be associated with neural tube defects. A pseudogene of this gene has been identified on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; Inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase
Gene Aliases: ITPK1; ITRPK1
UniProt ID: (Human) Q13572
Entrez Gene ID: (Human) 3705
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support