Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha v integrin; antigen identified by monoclonal antibody L230; CD51; integrin alpha V; Integrin alpha-V; integrin alphaVbeta3; integrin beta 6; Integrin beta-6; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 6; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); Vitronectin receptor subunit alpha
Gene Aliases: 1110004F14Rik; 2210409C20Rik; 2610028E01Rik; 4831415H04Rik; AI1H; CD51; D430040G12Rik; ITGAV; ITGB6; MSK8; VNRA; VTNR
UniProt ID: (Human) P06756, (Human) P18564, (Mouse) P43406, (Mouse) Q9Z0T9
Entrez Gene ID: (Human) 3685, (Human) 3694, (Mouse) 16410, (Mouse) 16420
Molecular Function: integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support