Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSQELVTLN VGGKIFTTRF STIKQFPASR LARMLDGRDQ EFKMVGGQIF VDRDGDLFSF ILDFLRTHQL LLPTEFSDYL RLQREALFYE LRSLVDLLNP YLLQPRPALV EVHFLSRNTQ AFFRVFGSCS KTIEMLTGRI TVFTEQPSAP TWNGNFFPPQ MTLLPLPPQR PSYHDLVFQC GSDSTTDNQT GVRYVSIKPD NRKLANGTNV LGLLIDTLLK EGFHLVSTRT VSSEDKTECY SFERIKSPEV LITNETPKPE TIIIPEQSQI KK
Inhibits potassium fluxes in cells. May regulate Kv1 family channel proteins by retaining a fraction of channels in endomembranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Potassium channel regulator; Potassium channel regulatory protein; Protein CLLD4; putative potassium channel regulatory protein
Gene Aliases: CLLD4; DLTET; KCNRG
UniProt ID: (Human) Q8N5I3
Entrez Gene ID: (Human) 283518
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support