Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPTWGARPAS PDRFAVSAEA ENKVREQQPH VERIFSVGVS VLPKDCPDNP HIWLQLEGPK ENASRAKEYL KGLCSPELQD EIHYPPKLHC IFLGAQGFFL DCLAWSTSAH LVPRAPGSLM ISGLTEAFVM AQSRVEELAE RLSWDFTPGP SSGASQCTGV LRDFSALLQS PGDAHREALL QLPLAVQEEL LSLVQEASSG QGPGALASWE GRSSALLGAQ CQGVRAPPSD GRESLDTGSM GPGDCRGARG DTYAVEKEGG KQGGPREMDL GWKELPGEEA WEREVALRPQ SVGGGARESA PLKGKALGKE EIALGGGGFC VHREPPGAHG SCHRAAQSRG ASLLQRLHNG NASPPRVPSP PPAPEPPWHC GDRGDCGDRG DVGDRGDKQQ GMARGRGPQW KRGARGGNLV TGTQRFKEAL QDPFTLCLAN VPGQPDLRHI VIDGSNVAMV HGLQHYFSSR GIAIAVQYFW DRGHRDITVF VPQWRFSKDA KVRESHFLQK LYSLSLLSLT PSRVMDGKRI SSYDDRFMVK LAEETDGIIV SNDQFRDLAE ESEKWMAIIR ERLLPFTFVG NLFMVPDDPL GRNGPTLDEF LKKPARTQGS SKAQHPSRGF AEHGKQQQGR EEEKGSGGIR KTRETERLRR QLLEVFWGQD HKVDFILQRE PYCRDINQLS EALLSLNF
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KH and NYN domain-containing protein; Protein KHNYN
Gene Aliases: KHNYN; KIAA0323
UniProt ID: (Human) O15037
Entrez Gene ID: (Human) 23351
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support