Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in many species including rat, mouse.
Specificity of this antibody: Krueppel-like factor 1, 2, 4, 5, 7, 12 (KLF1, KLF2, KLF4, KLF5, KLF7, KLF12).
The Kruppel family of transcription factors (KLFs) are a set of C2H2-type zinc-finger DNA binding proteins that regulate gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (intestinal Kruppel-like factor; AP-2 repressor; AP-2rep transcription factor; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; Basic transcription element-binding protein 2; BTE-binding protein 2; Colon krueppel-like factor; colon kruppel-like factor; EKLF; endothelial Kruppel-like zinc finger protein; Epithelial zinc finger protein EZF; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; GC box binding protein 2; GC-box-binding protein 2; gut Kruppel-like factor; Gut-enriched krueppel-like factor; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; KLF12 zinc finger transcriptional repressor; Klf5C isoform; Krueppel-like factor 1; Krueppel-like factor 12; Krueppel-like factor 2; Krueppel-like factor 4; Krueppel-like factor 5; Krueppel-like factor 7; Kruppel-like factor 1 (erythroid); Kruppel-like factor 12; Kruppel-like factor 2 (lung); Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); Kruppel-like factor 5 (intestinal); Kruppel-like factor 7 (ubiquitous); Kruppel-like factor LKLF; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor; monoclonal antibody A3D8; neonatal anemia; Transcription factor BTEB2; Transcriptional repressor AP-2rep; truncated EKLF; truncated erythroid Kruppel-like factor 1; Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor
Gene Aliases: 2700063E05Rik; 4930520J07Rik; 9830124P08Rik; AI225908; AI315654; AP-2rep; AP2REP; B130052C06Rik; BTEB2; CDAN4; CKLF; D530033K05Rik; EKLF; EZF; GKLF; HBFQTL6; HSPC122; IKLF; INLU; KLF1; KLF12; KLF2; KLF4; KLF5; KLF7; LKLF; Nan; UKLF; Zie
UniProt ID: (Human) Q13351, (Human) Q9Y4X4, (Human) Q9Y5W3, (Human) O43474, (Human) Q13887, (Human) O75840, (Mouse) O35738, (Mouse) Q60843, (Rat) Q9ET58, (Mouse) Q60793, (Mouse) Q9Z0Z7, (Mouse) Q99JB0
Entrez Gene ID: (Human) 10661, (Human) 11278, (Human) 10365, (Human) 9314, (Human) 688, (Human) 8609, (Mouse) 16596, (Rat) 304666, (Mouse) 16597, (Rat) 306110, (Mouse) 16598, (Rat) 306330, (Mouse) 16600, (Rat) 114505, (Rat) 84410, (Mouse) 12224, (Mouse) 93691, (Rat) 363243
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support