Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with rat and pig based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KSHIIID; Potassium voltage-gated channel subfamily C member 3; Voltage-gated potassium channel subunit Kv3.3
Gene Aliases: Kcnc3; Kcr2-3; KShIIID; Kv3.3
UniProt ID: (Mouse) Q63959
Entrez Gene ID: (Mouse) 16504
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support