Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGRAPEVGPS GPPLPSGLRD PECRPVDKDR KVCREPLADG PAPADAPREP CSAQDPHPQV STASSTLGPH SPEPLGVCRN PPPPCSPMLE ASDDEALLVC
LRP3 is a probable receptor, widely expressed, which may be involved in the internalization of lipophilic molecules and/or signal transduction. Its precise role is, however unclear, since it does not bind to very low density lipoprotein (VLDL) or to LRPAP1 in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 105 kDa low-density lipoprotein receptor-related protein; CTD-2540B15.13; hLRp105; low density lipoprotein receptor-related protein 3; Low-density lipoprotein receptor-related protein 3; LRP-3
Gene Aliases: LRP3
UniProt ID: (Human) O75074
Entrez Gene ID: (Human) 4037
Molecular Function: apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support