Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCPPQAQAEV GPTMTEKAEM VCAPSPAPAP PPKPASPGPP QVEEVGHRGG SSPPRLPPGV PVISLGHSRP PGVAMPTTEL GTLRPPLLQL STLGTAPPTL ALHYHPHPFL NSVYIGPAGP FSIFPSSRLK RRPSHCELDL AEGHQPQKVA RRVFTNSRER WRQQNVNGAF AELRKLLPTH PPDRKLSKNE VLRLAMKYIG FLVRLLRDQA AALAAGPTPP GPRKRPVHRV PDDGARRGSG RRAEAAARSQ PAPPADPDGS PGGAARPIKM EQTALSPEVR
Lyl-1, TAL1 and TAL2 are part of a family of basic helix-loop-helix (bHLH) proteins implicated in T cell acute leukemia. TAL1, also designated SCL, is a serine phosphoprotein and basic helix-loop-helix transcription factor known to regulate embryonic hematopoiesis. TAL2 is a protein involved in T cell acute lymphoblastic leukemia through a chromosomal translocation involving TAL2 and T cell receptor β chain genes. TAL2 includes a helix-loop-helix protein dimerization and DNA-binding domain that is homologous to TAL1 and Lyl-1 proto-oncogenes. Lyl-1 (lymphoblastic leukemia-derived sequence 1) is a nuclear protein. Endogenous Lyl-1 exists in complex with E2α proteins. Lyl-1 and E2α protein can form heterodimeric complexes with distinctive DNA-binding properties in hematolymphoid cells. Lyl-1 is involved in a chromosomal aberration which causes a form of T cell acute lymphoblastic leukemia (T-ALL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa18; Class A basic helix-loop-helix protein 18; lymphoblastic leukemia associated hematopoiesis regulator 1; lymphoblastic leukemia derived sequence 1; Lymphoblastic leukemia-derived sequence 1; Protein lyl-1
Gene Aliases: BHLHA18; LYL1
UniProt ID: (Human) P12980
Entrez Gene ID: (Human) 4066
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support