Search Thermo Fisher Scientific
MA1-81579 detects CD6 in rat samples.
MA1-81579 has been successfully used in immunohistochemistry (frozen sections), immunoprecipitation and Flow cytometry applications.
The immunogen used to produce MA1-81579 is from rat thoracic duct lymphocytes.
CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
Protein Aliases: CD6 antigen, Tp120; FLJ44171; T-cell differentiation antigen CD6