Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAL, a nonglycosylated integral membrane proteolipid, has been identified as a major internal component of glycolipid-enriched membrane (GEM) microdomains. MAL is the first integral membrane proteolipid component to have been shown as a necessary element in the cellular apical sorting apparatus for newly synthesized proteins. In its steady state this highly hydrophobic proteolipid, resides predominantly in the apical zone of polarized epithelial cells. It has also been shown in itinerant protein cycling between the plasma membrane and endosomes and in other cells. This 17-kDa proteolipid contains four putative transmembrane domains. These domains along with its protein trafficking properties are defined by the specific signals delegated by its specific amino acid sequence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17 kDa myelin vesicular protein; mal T-cell differentiation protein; MVP17; MyD88-adapter-like; Myelin and lymphocyte protein; myelin and lymphocyte protein, T-cell differentiation protein; NS 3; T-lymphocyte maturation-associated protein
Gene Aliases: MAL; MALGENE; Mpv17; Mvp17; Vip17
UniProt ID: (Human) P21145, (Mouse) O09198, (Rat) Q64349
Entrez Gene ID: (Human) 4118, (Mouse) 17153, (Rat) 25263
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support