Recommended positive controls: rat brain.
For long term storage, aliquot and freeze at -20°C.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: claustrin; DKFZp686E1099; DKFZp686F1345; FLJ38954; MAP-1B; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B heavy chain; Microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Neuraxin; protein phosphatase 1, regulatory subunit 102
Gene Aliases: A230055D22; AI843217; FUTSCH; I79_010564; LC1; MAP1B; MAP5; Mtap-5; Mtap1b; Mtap5; PPP1R102
UniProt ID: (Human) P46821, (Rat) P15205, (Mouse) P14873
Entrez Gene ID: (Bovine) 514739, (Human) 4131, (Chicken) 396174, (Rat) 29456, (Chinese hamster) 100759518, (Mouse) 17755
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support