Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
MAP3K15 is a component of a protein kinase signal transduction cascade.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis signal-regulating kinase 3; MAPK/ERK kinase kinase 15; MEK kinase 15; MEKK 15; Mitogen-activated protein kinase kinase kinase 15
Gene Aliases: ASK3; bA723P2.3; MAP3K15
UniProt ID: (Human) Q6ZN16
Entrez Gene ID: (Human) 389840
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support