Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EVSEDAKQLI RLLLKTDPTE RLTITQFMNH PWINQSMVVP QTPLHTARVL QEDKDHWDEV KEEMTSALAT MRVDYDQVKI KDLKTSNNRL LNKRRKKQAG SSSASQGCNN Q
MAPKAPK3 functions as an integrative element of signaling in both mitogen and stress responses. MAPKAPK3 functions as a mitogen-activated protein kinase (MAP kinase)- activated protein kinase. MAP kinases are also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. MAPKAPK3 was shown to be activated by growth inducers and stress stimulation of cells. In vitro studies demonstrated that ERK, p38 MAP kinase and Jun N-terminal kinase were all able to phosphorylate and activate this kinase, which suggested the role of this kinase as an integrative element of signaling in both mitogen and stress responses. MAPKAPK3 was reported to interact with, phosphorylate and repress the activity of E47, which is a basic helix-loop-helix transcription factor known to be involved in the regulation of tissue-specific gene expression and cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3pK; Chromosome 3p kinase; MAP kinase-activated protein kinase 3; MAPK-activated protein kinase 3; MAPKAP kinase 3; MAPKAPK-3; mitogen activated protein kinase-activated protein kinase 3; OTTHUMP00000210806; OTTHUMP00000210807
Gene Aliases: 3PK; AI874665; MAPKAP-K3; MAPKAP3; MAPKAPK-3; MAPKAPK3; MapkKapk3; MK-3; MK3
UniProt ID: (Human) Q16644, (Mouse) Q3UMW7
Entrez Gene ID: (Human) 7867, (Mouse) 102626
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support