Product References
Vaccination with antigen-transfected, NKT cell ligand-loaded, human cells elicits robust in situ immune responses by dendritic cells.
Cancer research
Shimizu K,Mizuno T,Shinga J,Asakura M,Kakimi K,Ishii Y,Masuda K,Maeda T,Sugahara H,Sato Y,Matsushita H,Nishida K,Hanada K,Dorrie J,Schaft N,Bickham K,Koike H,Ando T,Nagai R,Fujii S
MA5-14017 was used in western blot to develop human artificial adjuvant vector cells for cancer immunotherapy
Tue Jan 01 00:00:00 EST 2013
Ambiguous melanocytic tumors in a tertiary referral center: the contribution of fluorescence in situ hybridization (FISH) to conventional histopathologic and immunophenotypic analyses.
The American journal of surgical pathology
Tetzlaff MT,Wang WL,Milless TL,Curry JL,Torres-Cabala CA,McLemore MS,Ivan D,Bassett RL,Prieto VG
MA5-14017 was used in immunohistochemistry to study the use of FISH as a diagnostic aid in patients with ambiguous melanocytic tumors
Sun Dec 01 00:00:00 EST 2013
Characterization of the bioactive motif of neuregulin-1, a fibroblast-derived paracrine factor that regulates the constitutive color and the function of melanocytes in human skin.
Pigment cell & melanoma research
Choi W,Kolbe L,Hearing VJ
MA5-14017 was used in western blot to study a peptide sequence motif within neuregulin-1 that promotes the biosynthesis of melanin but not melanocyte proliferation
Sun Jul 01 00:00:00 EDT 2012
Intraglomerular crescentic metastases of malignant melanoma.
Human pathology
Ozluk Y,Kilicaslan I,Kazancioglu R,Demirkesen C,Derin D,Mandel NM
MA5-14017 was used in immunohistochemistry to report on a case of intraglomerular crescentic metastases of malignant melanoma
Thu Dec 01 00:00:00 EST 2011
Expression of the embryological morphogen Nodal in stage III/IV melanoma.
Melanoma research
Hooijkaas AI,Gadiot J,van Boven H,Blank C
MA5-14017 was used in immunohistochemistry to investigate the role of Nodal during melanoma progression
Thu Dec 01 00:00:00 EST 2011
Regulation of human skin pigmentation in situ by repetitive UV exposure: molecular characterization of responses to UVA and/or UVB.
The Journal of investigative dermatology
Choi W,Miyamura Y,Wolber R,Smuda C,Reinhold W,Liu H,Kolbe L,Hearing VJ
MA5-14017 was used in immunohistochemistry to study the molecular responses of the skin pigmentation system to repeated exposure to UVA and UVB
Tue Jun 01 00:00:00 EDT 2010
Glycoprotein nonmetastatic melanoma protein b, a melanocytic cell marker, is a melanosome-specific and proteolytically released protein.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hoashi T,Sato S,Yamaguchi Y,Passeron T,Tamaki K,Hearing VJ
MA5-14017 was used in western blot to study the proteolytic release of glycoprotein nonmetastatic melanoma protein b from melanosomes
Sat May 01 00:00:00 EDT 2010
Cellular blue nevus of the uterus: a case report and review of the literature.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Eskue K,Prieto VG,Malpica A
MA5-14017 was used in immunohistochemistry to report on a case of cellular blue nevus of the uterus
Mon Nov 01 00:00:00 EDT 2010
Vaccine-specific local T cell reactivity in immunotherapy-associated vitiligo in melanoma patients.
Cancer immunology, immunotherapy : CII
Jacobs JF,Aarntzen EH,Sibelt LA,Blokx WA,Boullart AC,Gerritsen MJ,Hoogerbrugge PM,Figdor CG,Adema GJ,Punt CJ,de Vries IJ
MA5-14017 was used in immunohistochemistry to investigate the association between vitiligo and immunetherapy for melanoma
Thu Jan 01 00:00:00 EST 2009
Diagnostic challenges of metastatic spindle cell melanoma on fine-needle aspiration specimens.
Cancer
Piao Y,Guo M,Gong Y
MA5-14017 was used in immunohistochemistry to review the diagnostic difficulty of metastatic spindle cell melanoma
Fri Apr 25 00:00:00 EDT 2008
Glomus tumor and glomangioma of the nerve. Report of two cases.
Journal of neurosurgery
Scheithauer BW,Rodriguez FJ,Spinner RJ,Dyck PJ,Salem A,Edelman FL,Amrami KK,Fu YS
MA5-14017 was used in immunohistochemistry to report on two cases of glomus tumor and glomangioma of the nerve
Fri Feb 01 00:00:00 EST 2008
Cyclobutane pyrimidine dimer formation and p53 production in human skin after repeated UV irradiation.
Experimental dermatology
Yamaguchi Y,Coelho SG,Zmudzka BZ,Takahashi K,Beer JZ,Hearing VJ,Miller SA
MA5-14017 was used in immunocytochemistry to study the potential protective effects of facultative pigmentation following repeated exposure to UV irradiation
Sat Nov 01 00:00:00 EDT 2008
Dendritic cell vaccine with mRNA targeted to the proteasome by polyubiquitination.
Biochemical and biophysical research communications
Hosoi A,Takeda Y,Sakuta K,Ueha S,Kurachi M,Kimura K,Maekawa R,Kakimi K
MA5-14017 was used in western blot to study the therapeutic potential of a dendritic cell vaccine using mRNAs encoding tumor-associated antigens targeted to the proteasome
Fri Jun 27 00:00:00 EDT 2008
McCune-Albright syndrome in a boy may present with a monolateral macroorchidism as an early and isolated clinical manifestation.
Hormone research
Arrigo T,Pirazzoli P,De Sanctis L,Leone O,Wasniewska M,Messina MF,De Luca F
MA5-14017 was used in immunohistochemistry to report on a case of McCune-Albright syndrome with monolateral macroorchidism
Wed Aug 16 00:00:00 EDT 2006
Immunogenicity, including vitiligo, and feasibility of vaccination with autologous GM-CSF-transduced tumor cells in metastatic melanoma patients.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Luiten RM,Kueter EW,Mooi W,Gallee MP,Rankin EM,Gerritsen WR,Clift SM,Nooijen WJ,Weder P,van de Kasteele WF,Sein J,van den Berk PC,Nieweg OE,Berns AM,Spits H,de Gast GC
MA5-14017 was used in immunohistochemistry to study the therapeutic potential of autologous tumor cells virally transduced with GM-CSF in metastatic melanoma patients
Sat Dec 10 00:00:00 EST 2005
Mechanisms of skin tanning in different racial/ethnic groups in response to ultraviolet radiation.
The Journal of investigative dermatology
Tadokoro T,Yamaguchi Y,Batzer J,Coelho SG,Zmudzka BZ,Miller SA,Wolber R,Beer JZ,Hearing VJ
MA5-14017 was used in immunohistochemistry to study the mechanisms by which UV radiation stimulates skin pigmentation in different ethnic groups
Wed Jun 01 00:00:00 EDT 2005
MART-1 is required for the function of the melanosomal matrix protein PMEL17/GP100 and the maturation of melanosomes.
The Journal of biological chemistry
Hoashi T,Watabe H,Muller J,Yamaguchi Y,Vieira WD,Hearing VJ
MA5-14017 was used in immunoprecipitation to study the role of MART-1 on PMEL17/GP100 function and melanosome maturation
Fri Apr 08 00:00:00 EDT 2005
On the role of melanoma-specific CD8+ T-cell immunity in disease progression of advanced-stage melanoma patients.
Clinical cancer research : an official journal of the American Association for Cancer Research
van Oijen M,Bins A,Elias S,Sein J,Weder P,de Gast G,Mallo H,Gallee M,Van Tinteren H,Schumacher T,Haanen J
MA5-14017 was used in immunohistochemistry to study the role of CD8+ T-cell immunity in disease progression of advanced-stage melanoma
Thu Jul 15 00:00:00 EDT 2004
Magnetic bead RT-PCR: establishment of a new method for detecting circulating melanoma cells.
Melanoma research
Georgieva J,Milling A,Orfanos CE,Geilen CC
MA5-14017 was used in flow cytometry and immunoprecipitation to develop a novel magnetic-activated cell separation/RT-PCR method for detecting circulating melanoma cell anaylsis
Thu Aug 01 00:00:00 EDT 2002