Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLAAAAEPG AGSQHLEVRD EVAEKCQKLF LDFLEEFQSS DGEIKYLQLA EELIRPERNT LVVSFVDLEQ FNQQLSTTIQ EEFYRVYPYL CRALKTFVKD RKEIPLAKDF YVAFQDLPTR HKIRELTSSR IGLLTRISGQ VVRTHPVHPE LVSGTFLCLD CQTVIRDVEQ QFKYTQPNIC RNPVCANRRR FLLDTNKSRF VDFQKVRIQE TQAELPRGSI PRSLEVILRA EAVESAQAGD KCDFTGTLIV VPDVSKLSTP GARAETNSRV SGVDGYETEG IRGLRALGVR DLSYRLVFLA CCVAPTNPRF GGKELRDEEQ TAESIKNQMT VKEWEKVFEM SQDKNLYHNL CTSLFPTIHG NDEVKRGVLL MLFGGVPKTT GEGTSLRGDI NVCIVGDPST AKSQFLKHVE EFSPRAVYTS GKASSAAGLT AAVVRDEESH EFVIEAGALM LADNGVCCID EFDKMDVRDQ VAIHEAMEQQ TISITKAGVK ATLNARTSIL AAANPISGHY DRSKSLKQNI NLSAPIMSRF DLFFILVDEC NEVTDYAIAR RIVDLHSRIE ESIDRVYSLD DIRRYLLFAR QFKPKISKES EDFIVEQYKH LRQRDGSGVT KSSWRITVRQ LESMIRLSEA MARMHCCDEV QPKHVKEAFR LLNKSIIRVE TPDVNLDQEE EIQMEVDEGA GGINGHADSP APVNGINGYN EDINQESAPK ASLRLGFSEY CRISNLIVLH LRKVEEEEDE SALKRSELVN WYLKEIESEI DSEEELINKK RIIEKVIHRL THYDHVLIEL TQAGLKGSTE GSESYEEDPY LVVNPNYLLE D
MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA replication licensing factor MCM6; MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); minichromosome maintenance deficient (mis5, S. pombe) 6; p105MCM
Gene Aliases: MCG40308; MCM6; Mis5; P105MCM
UniProt ID: (Human) Q14566
Entrez Gene ID: (Human) 4175
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support