Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAAAVAMETD DAGNRLRFQL ELEFVQCLAN PNYLNFLAQR GYFKDKAFVN YLKYLLYWKD PEYAKYLKYP QCLHMLELLQ YEHFRKELVN AQCAKFIDEQ QILHWQHYSR KRMRLQQALA EQQQQNNTSG K
MED31 is the component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27436; FLJ36714; hSOH1; Mediator complex subunit 31; Mediator complex subunit SOH1; Mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription, subunit 31 homolog
Gene Aliases: 3110004H13Rik; CGI-125; MED31; SOH1
UniProt ID: (Human) Q9Y3C7
Entrez Gene ID: (Human) 51003
Molecular Function: RNA metabolism protein nucleic acid metabolism protein general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support