Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein Meis1; leukemogenic homolog protein; Meis1, myeloid ecotropic viral integration site 1 homolog; MGC43380; WUGSC:H_NH0444B04.1
Gene Aliases: MEIS1
UniProt ID: (Human) O00470
Entrez Gene ID: (Human) 4211
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support