Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody detects the pro form of human or mouse Meprin beta Subunit/MEP1B in Western blots. This antibody does not react with the mature form.
Reconstitute at 0.5 mg/mL in sterile PBS.
Meprins are multidomain zinc metalloproteases that are highly expressed in mammalian kidney and intestinal brush border membranes, and in leukocytes and certain cancer cells. They are involved in the hydrolysis of a variety of peptide and protein substrates, and have been implicated in cancer and intestinal inflammation. Mature meprins are oligomers of evolutionarily related, but separately encoded alpha and/or beta subunits. Homooligomers of alpha subunit are secreted, whereas, oligomers containing the beta subunit are plasma membrane-bound. This gene encodes the beta subunit. Targeted disruption of this gene in mice affects embryonic viability, renal gene expression profiles, and distribution of the membrane-associated alpha subunit in kidney and intestine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.4.24.63; Endopeptidase-2; meprin A; Meprin A subunit beta; meprin A, beta; Meprin B; meprin beta; N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta; PABA peptide hydrolase; PPH beta
Gene Aliases: Mep-1b; MEP1B
UniProt ID: (Human) Q16820, (Mouse) Q61847
Entrez Gene ID: (Human) 4225, (Mouse) 17288
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support