Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; hMETTL3; m6a methyltransferase; methyltransferase-like 3; Methyltransferase-like protein 3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3
Gene Aliases: 2310024F18Rik; IME4; M6A; METTL3; MT-A70; MTA70; Spo8
UniProt ID: (Human) Q86U44, (Mouse) Q8C3P7
Entrez Gene ID: (Human) 56339, (Mouse) 56335
Molecular Function: RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support