Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Mouse anti Bovine MHC Class II Monomorphic antibody, clone IL-A21 recognizes a monomorphic epitope within the bovine MHC II cell surface antigen.
MHC II Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: BoLA-DR-alpha; BoLA-DRB3.2; DR beta-chain antigen binding domain; DR-beta chain MHC class II; histocompatibility complex, class II, DR beta 3; leukocyte antigen DRB3; major histocompatibility complex class II DR-beta chain; major histocompatibility complex, class II, DR beta 3; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen BoLA-DRB3; MHC class II DQA; MHC class II DR beta chain; MHC class II DRB3; MHC class II leukocyte antigen BoLA-DRB3; MHC(BoLA) class II DR-beta chain
Gene Aliases: BLA-DRB3; BoLA-DQA; BoLA-DQA1; BoLA-DRB; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; DRB3; LA-DRB
Entrez Gene ID: (Bovine) 282534, (Bovine) 282535, (Bovine) 282536, (Bovine) 282494, (Bovine) 282495, (Bovine) 506214, (Bovine) 282530
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support