Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSLIP1; MIF4G domain-containing protein; SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1
Gene Aliases: AD023; MIF4GD; MIFD; SLIP1
UniProt ID: (Human) A9UHW6
Entrez Gene ID: (Human) 57409
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support