Product References
Empagliflozin mitigates type 2 diabetes-associated peripheral neuropathy: a glucose-independent effect through AMPK signaling.
Archives of pharmacal research
Abdelkader NF,Elbaset MA,Moustafa PE,Ibrahim SM
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Fri Jul 01 00:00:00 EDT 2022
Empagliflozin mitigates type 2 diabetes-associated peripheral neuropathy: a glucose-independent effect through AMPK signaling.
Archives of pharmacal research
Abdelkader NF,Elbaset MA,Moustafa PE,Ibrahim SM
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Fri Jul 01 00:00:00 EDT 2022
Predictive Significance of Two MMP-9 Promoter Polymorphisms and Acetylated c-Jun Transcription Factor for Papillary Thyroid Carcinoma Advancement.
Diagnostics (Basel, Switzerland)
Rončević J,Janković Miljuš J,Išić Denčić T,Božić V,Živaljević V,Šelemetjev S,Đorić I
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Fri Aug 12 00:00:00 EDT 2022
In Vitro Evaluation of the Anti-Inflammatory Effect of KMUP-1 and In Vivo Analysis of Its Therapeutic Potential in Osteoarthritis.
Biomedicines
Huang SE,Sulistyowati E,Chao YY,Wu BN,Dai ZK,Hsu JH,Yeh JL
MA5-14228 was used in Western Blot to investigate the in vitro anti-inflammatory and in vivo anti-osteoarthritis effects of KMUP-1.
Fri May 28 00:00:00 EDT 2021
In Vitro Evaluation of the Anti-Inflammatory Effect of KMUP-1 and In Vivo Analysis of Its Therapeutic Potential in Osteoarthritis.
Biomedicines
Huang SE,Sulistyowati E,Chao YY,Wu BN,Dai ZK,Hsu JH,Yeh JL
MA5-14228 was used in Western Blot to investigate the in vitro anti-inflammatory and in vivo anti-osteoarthritis effects of KMUP-1.
Fri May 28 00:00:00 EDT 2021
Spatio-Temporal Expression Pattern of Ki-67, pRB, MMP-9 and Bax in Human Secondary Palate Development.
Life (Basel, Switzerland)
Šimić Bilandžija T,Vukojević K,Ćorić A,Vuković Kekez I,Medvedec Mikić I,Lasić Arapović L,Filipović N,Anđelić J,Saraga-Babić M,Kalibović Govorko D
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunocytochemistry
Sat Feb 20 00:00:00 EST 2021
TAGLN and High-mobility Group AT-Hook 2 (HMGA2) Complex Regulates TGF-β-induced Colorectal Cancer Metastasis.
OncoTargets and therapy
Zhou H,Li L,Xie W,Wu L,Lin Y,He X
MA5-14228 was used in Western Blotting to investigate the role of transgelin and high-mobility group AT-hook 2 in the progression of colon cancer.
Sun Apr 17 00:00:00 EDT 2022
TAGLN and High-mobility Group AT-Hook 2 (HMGA2) Complex Regulates TGF-β-induced Colorectal Cancer Metastasis.
OncoTargets and therapy
Zhou H,Li L,Xie W,Wu L,Lin Y,He X
MA5-14228 was used in Western Blotting to investigate the role of transgelin and high-mobility group AT-hook 2 in the progression of colon cancer.
Sun Apr 17 00:00:00 EDT 2022
Disturbed balance in the expression of MMP9 and TIMP3 in cerebral amyloid angiopathy-related intracerebral haemorrhage.
Acta neuropathologica communications
Jäkel L,Kuiperij HB,Gerding LP,Custers EEM,van den Berg E,Jolink WMT,Schreuder FHBM,Küsters B,Klijn CJM,Verbeek MM
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Mon Jul 06 00:00:00 EDT 2020
Disturbed balance in the expression of MMP9 and TIMP3 in cerebral amyloid angiopathy-related intracerebral haemorrhage.
Acta neuropathologica communications
Jäkel L,Kuiperij HB,Gerding LP,Custers EEM,van den Berg E,Jolink WMT,Schreuder FHBM,Küsters B,Klijn CJM,Verbeek MM
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Mon Jul 06 00:00:00 EDT 2020
The beneficial influence of rhubarb on 5-fluorouracil-induced ileal mucositis and the combined role of aquaporin-4, tumour necrosis factor-α, nuclear factor-kappa B & matrix metalloproteinase-9 in rat model: histological study.
Anatomy & cell biology
Kamar SS,Baky MH,Omar AI
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Tue Jun 30 00:00:00 EDT 2020
Astrocyte-derived VEGF increases cerebral microvascular permeability under high salt conditions.
Aging
Deng Z,Zhou L,Wang Y,Liao S,Huang Y,Shan Y,Tan S,Zeng Q,Peng L,Huang H,Lu Z
MA5-14228 was used in Western Blotting to reveal a new axis linking a high salt diet (HSD) to increased cerebral microvascular permeability through a VEGF-initiated inflammatory response.
Mon Jun 22 00:00:00 EDT 2020
A Novel Role of Irbesartan in Gastroprotection against Indomethacin-Induced Gastric Injury in Rats: Targeting DDAH/ADMA and EGFR/ERK Signaling.
Scientific reports
Shahin NN,Abdelkader NF,Safar MM
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Fri Mar 09 00:00:00 EST 2018
The pattern of CD44 and matrix metalloproteinase 9 expression is a useful predictor of ulcerative colitis-associated dysplasia and neoplasia.
Annals of diagnostic pathology
AbdElazeem MA,El-Sayed M
MA5-14228 was used in immunohistochemistry - paraffin section to examine CD44 and MMP-9 expression in different ulcerative colitis lesions
Tue Dec 01 00:00:00 EST 2015
Recruitment of Matrix Metalloproteinase-9 (MMP-9) to the Fibroblast Cell Surface by Lysyl Hydroxylase 3 (LH3) Triggers Transforming Growth Factor-β (TGF-β) Activation and Fibroblast Differentiation.
The Journal of biological chemistry
Dayer C,Stamenkovic I
MA5-14228 was used in immunocytochemistry and western blot to study the recruitment of MMP-9 to cell surface and its effect on fibroblast differentiation
Fri May 29 00:00:00 EDT 2015
In vivo effects of zoledronic acid on oral mucosal epithelial cells.
Oral diseases
Allam E,Allen M,Chu TM,Ghoneima A,Jack Windsor L
MA5-14228 was used in immunohistochemistry to investigate the influence of zoledronic acid on oral mucosal epithelial cells in a dog model
Fri Apr 01 00:00:00 EDT 2011
Noninvasive limb ischemic preconditioning protects against myocardial I/R injury in rats.
The Journal of surgical research
Li SJ,Wu YN,Kang Y,Yin YQ,Gao WZ,Liu YX,Lou JS
MA5-14228 was used in immunohistochemistry to compare the protective effect of noninvasive limb ischemia preconditioning against myocardial ischemia/reperfusion injury in rats
Mon Nov 01 00:00:00 EDT 2010
Human mammary fibroblasts stimulate invasion of breast cancer cells in a three-dimensional culture and increase stroma development in mouse xenografts.
BMC cancer
Olsen CJ,Moreira J,Lukanidin EM,Ambartsumian NS
MA5-14228 was used in western blot to study the effects of fibroblasts on mammary carcinoma cells.
Thu Aug 19 00:00:00 EDT 2010
Cardosins improve neuronal regeneration after cell disruption: a comparative expression study.
Cell biology and toxicology
Duarte AS,Duarte EP,Correia A,Pires E,Barros MT
MA5-14228 was used in western blot to study the use of the plant enzyme cardosin for tissue disaggregation and isolation of rat embryonic neurons
Wed Apr 01 00:00:00 EDT 2009
Biological and clinical relevance of matrix metalloproteinases 2 and 9 in acute myeloid leukaemias and myelodysplastic syndromes.
European journal of haematology
Travaglino E,Benatti C,Malcovati L,Della Porta MG,Gallì A,Bonetti E,Rosti V,Cazzola M,Invernizzi R
MA5-14228 was used in immunocytochemistry to study the biological and clinical relevance of MMP2 and MMP9 in acute myeloid leukaemias and myelodysplastic syndromes
Sat Mar 01 00:00:00 EST 2008
Impaired leukocyte influx in cervix of postterm women not responding to prostaglandin priming.
Reproductive biology and endocrinology : RB&E
Sahlin L,Stjernholm-Vladic Y,Roos N,Masironi B,Ekman-Ordeberg G
MA5-14228 was used in immunohistochemistry to investigate the mechanism for the non-responsiveness to prostaglandin priming in cervix of postterm women
Tue Sep 02 00:00:00 EDT 2008
Pathogenic sequence for dissecting aneurysm formation in a hypomorphic polycystic kidney disease 1 mouse model.
Arteriosclerosis, thrombosis, and vascular biology
Hassane S,Claij N,Lantinga-van Leeuwen IS,Van Munsteren JC,Van Lent N,Hanemaaijer R,Breuning MH,Peters DJ,DeRuiter MC
MA5-14228 was used in immunohistochemistry to study vascular remodeling in a hypomorphic Pkd1(nl/nL) mouse model with dissecting aneurysms in the aorta
Mon Oct 01 00:00:00 EDT 2007
Improved surgical mesh integration into the rat abdominal wall with arginine administration.
Biomaterials
Arbos MA,Ferrando JM,Quiles MT,Vidal J,López-Cano M,Gil J,Manero JM,Peña J,Huguet P,Schwartz-Riera S,Reventós J,Armengol M
MA5-14228 was used in western blot to study the role of arginine administration in improving surgical mesh integration into the rat abdominal wall
Wed Feb 01 00:00:00 EST 2006
Simvastatin suppresses experimental aortic aneurysm expansion.
Journal of vascular surgery
Kalyanasundaram A,Elmore JR,Manazer JR,Golden A,Franklin DP,Galt SW,Zakhary EM,Carey DJ
MA5-14228 was used in western blot to study the therapeutic benefits of simvastatin against experimental aortic aneurysm expansion
Sun Jan 01 00:00:00 EST 2006
Time course of matrix metalloproteases and tissue inhibitors in bleomycin-induced pulmonary fibrosis.
European journal of histochemistry : EJH
Oggionni T,Morbini P,Inghilleri S,Palladini G,Tozzi R,Vitulo P,Fenoglio C,Perlini S,Pozzi E
MA5-14228 was used in immunohistochemistry to investigate the distribution, expression and activity of matrix metalloproteases and tissue inhibitors during extracellular matrix remodeling in pulmonary fibrosis
Tue Mar 13 00:00:00 EDT 2007
Expression and localisation of matrix metalloproteinases and their natural inhibitors in fistulae of patients with Crohn's disease.
Gut
Kirkegaard T,Hansen A,Bruun E,Brynskov J
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Sat May 01 00:00:00 EDT 2004
Expression and localisation of matrix metalloproteinases and their natural inhibitors in fistulae of patients with Crohn's disease.
Gut
Kirkegaard T,Hansen A,Bruun E,Brynskov J
Published figure using MMP9 monoclonal antibody (Product # MA5-14228) in Immunohistochemistry
Sat May 01 00:00:00 EDT 2004
Effect of gamma-synuclein overexpression on matrix metalloproteinases in retinoblastoma Y79 cells.
Archives of biochemistry and biophysics
Surgucheva IG,Sivak JM,Fini ME,Palazzo RE,Surguchov AP
MA5-14228 was used in western blot to study the effect of gamma-synuclein overexpression on MMPs in retinoblastoma cells
Sat Feb 01 00:00:00 EST 2003