Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Esa1p-associated factor 3 homolog; MGC10631; MORF-related gene 15 protein; MORF-related gene on chromosome 15; Mortality factor 4-like protein 1; Protein MSL3-1; testis expressed gene 189; Testis-expressed gene 189 protein; Transcription factor-like protein MRG15
Gene Aliases: Eaf3; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; mKIAA4002; MORF4L1; MORFRG15; MRG15; PP368; S863-6; TEG-189; Tex189
UniProt ID: (Human) Q9UBU8, (Rat) Q6AYU1, (Mouse) P60762
Entrez Gene ID: (Human) 10933, (Rat) 300891, (Mouse) 21761
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support