Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
N-Myc protein is member of the MYC family of transcription factor proteins with a basic helix-loop-helix (bHLH) domain. N-Myc is similarly regulated and functionally complementary to C-Myc in the context of various cellular growth and differentiation processes. MYCN gene is considered the hallmark of neuroblastoma, amplification and overexpression of the MYCN proto-oncogene occurs in approximately 20% of neuroblastomas and is associated with advanced stage disease, rapid tumor progression, and poor prognosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Avian myelocytomatosis viral (v-myc) related oncogene neuroblastoma derived (Nmyc); Avian myelocytomatosis viral (v-myc) related oncogene, neuroblastoma derived (Nmyc); bHLHe37; Class E basic helix-loop-helix protein 37; N-myc proto-oncogene protein; neuroblastoma MYC oncogene; neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene; oncogene NMYC; oncogene pp65/67; pp65/67; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived
Gene Aliases: BHLHE37; MODED; MYCN; N-myc; Nmuc1; NMYC; Nmyc1; ODED
UniProt ID: (Human) P04198, (Rat) Q63379
Entrez Gene ID: (Human) 4613, (Rat) 298894
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support