Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 100 µL of distilled water.
Centrifuge to remove any insoluble material.
Expressed in the cochlea; particularly in the inner and outer hair cells. Defects in Mcoln3 are the cause of the varitin-waddler (Va) phenotype. Classical Va mice exhibit early-onset hearing loss, vestibular defects, pigmentation abnormalities and perinatal lethality. The phenotype varitin-waddler Jackcon (Va-J), which arose in a cross segregating for Va, is similar but less severe. Belongs to the transient receptor family, polycystin subfamily.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ11006; FLJ36629; MCLN3; Mcoln3 mucolipin 3; MGC124245; MGC124246; MGC71509; Mucolipin-3; Transient receptor potential channel mucolipin 3; TRPML3; varitint-waddler
Gene Aliases: Mcoln3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support