Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetyltransferase 2; ATase2; Camello-like protein 1; CCNAT; Cysteinyl-conjugate N-acetyltransferase; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); NAT8; probable N-acetyltransferase 8
Gene Aliases: ATase2; CCNAT; CML1; GLA; Hcml1; NAT8; TSC501; TSC510
UniProt ID: (Human) Q9UHE5
Entrez Gene ID: (Human) 9027
Molecular Function: acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support