Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ30225; FLJ34463; I-kappa-B-zeta; Ikappa B-zeta variant 3; IkappaB-zeta; ikappaBzeta; IkB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; MAIL; Molecule possessing ankyrin repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340
Gene Aliases: IKBZ; INAP; MAIL; NFKBIZ
UniProt ID: (Human) Q9BYH8
Entrez Gene ID: (Human) 64332
Molecular Function: protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support