Search Thermo Fisher Scientific
The MA1-7429 antibody reacts with Neisseria gonorrhoeae porin in bacterial and infected tissue samples.
MA1-7429 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7429 antibody was raised against N. gonorrhoeae Porin.