Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Predicted reactivity: Mouse (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20363; HGNC:16395; ORP-10; OSBP-related protein 10; OTTHUMP00000161257; oxysterol binding protein-like 10; Oxysterol-binding protein-related protein 10
Gene Aliases: ORP10; OSBP9; OSBPL10
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support