Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: MOR245-7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GA_x6K02T2N82Q-3465-3764; GA_x6K02T2Q125-72882187-72881249; MOR245-28_p; olfactory receptor; olfactory receptor 1303; olfactory receptor 276, pseudogene 1; olfactory receptor MOR245-7
Gene Aliases: MOR245-7; Olfr276; Olfr276-ps1
Entrez Gene ID: (Mouse) 258397
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support