Search Thermo Fisher Scientific
In Western blot applications, this antibody detects a band at ~40kDa, which represents the pro-enzyme of DNase II. A suggested positive control is human spleen tissue lysate.
PA5-19900 can be used with blocking peptide PEP-0026.
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codominant alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.
Protein Aliases: Acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; Deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; Lysosomal DNase II; R31240_2
Gene Aliases: DNASE2; DNASE2A; DNL; DNL2
UniProt ID: (Human) O00115
Entrez Gene ID: (Human) 1777
Molecular Function: endodeoxyribonuclease