Search Thermo Fisher Scientific
Predicted to react with bovine and mouse based on sequence homology.
This gene encodes a transmembrane protein that interacts with the beta subunit of a sodium/potassium-transporting ATPase. A chromosomal translocation involving this gene is a cause of lymphoma. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2014].
Protein Aliases: Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 2; Protein FAM77B; Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 2; T-cell lymphoma breakpoint-associated target protein 1
Gene Aliases: FAM77B; NKAIN2; TCBA1