Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: SRLLRSDFCP EHVPKKQHKD KFLTE
Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Calcium channel facilitator that increases calcium flux by generating a larger window current and slowing inactivation of the L-type CACNA1C/CaV1.2 channel. Regulation of intracellular calcium by Calfacilitin is required for neural plate formation.
Protein Aliases: Calfacilitin; TLC domain-containing protein 1
Gene Aliases: TLCD1